Learn More
Hampton Research PEG/ION SCREEN NO21 185ML
SDP
Supplier:  Hampton Research HR292221
Encompass
HR2-922-21/PEG/Ion Screen #21 (B9), 185 ml - 0.2 M Sodium formate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-184-4