Learn More
Hampton Research PEG/ION SCREEN NO26 185ML
SDP
Supplier:  Hampton Research HR292226
Encompass
HR2-922-26/PEG/Ion Screen #26 (C2), 185 ml - 0.2 M Zinc acetate dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-184-9