Learn More
Hampton Research PEG/ION SCREEN NO28 185ML
SDP
Supplier:  Hampton Research HR292228
Encompass
HR2-922-28/PEG/Ion Screen #28 (C4), 185 ml - 0.2 M Calcium acetate hydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-185-1