Learn More
Hampton Research PEG/ION SCREEN NO30 185ML
SDP
Supplier:  Hampton Research HR292230
Encompass
HR2-922-30/PEG/Ion Screen #30 (C6), 185 ml - 0.2 M Ammonium acetate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-185-3