Learn More
Hampton Research PEG/ION SCREEN NO31 185ML
SDP
Supplier:  Hampton Research HR292231
Encompass
HR2-922-31/PEG/Ion Screen #31 (C7), 185 ml - 0.2 M Lithium sulfate monohydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-185-4