Learn More
Hampton Research PEG/ION SCREEN NO32 185ML
SDP
Supplier:  Hampton Research HR292232
Encompass
HR2-922-32/PEG/Ion Screen #32 (C8), 185 ml - 0.2 M Magnesium sulfate heptahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-185-5