Learn More
Hampton Research PEG/ION SCREEN NO34 185ML
SDP
Supplier:  Hampton Research HR292234
Encompass
HR2-922-34/PEG/Ion Screen #34 (C10), 185 ml - 0.2 M Potassium sulfate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-185-7