Learn More
Hampton Research PEG/ION SCREEN NO43 185ML
SDP
Supplier:  Hampton Research HR292243
Encompass
HR2-922-43/PEG/Ion Screen #43 (D7), 185 ml - 0.2 M Ammonium phosphate monobasic, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-186-6