Learn More
Hampton Research PEG/ION SCREEN NO44 185ML
SDP
Supplier:  Hampton Research HR292244
Encompass
HR2-922-44/PEG/Ion Screen #44 (D8), 185 ml - 0.2 M Ammonium phosphate dibasic, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-186-7