Learn More
Hampton Research PEG/ION SCREEN NO47 185ML
SDP
Supplier:  Hampton Research HR292247
Encompass
HR2-922-47/PEG/Ion Screen #47 (D11), 185 ml - 0.2 M Potassium citrate tribasic monohydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-187-0