Learn More
BEZWADA BIOMEDICAL LLC PEG Dimethacrylate, MW 2000, 5.0 GM
SDP
Supplier:  BEZWADA BIOMEDICAL LLC 72K001
Encompass_Preferred
Acrylate/Dimethacrylate end functionalization enables crosslinking. Polymers used in drug delivery and various biomedical applications
Catalog No. 50-190-4502