Learn More
BEZWADA BIOMEDICAL LLC Poly (PEG-co-L-lactide-co-glycolde), PEG:L-lactide:glycolide, 5.0 GM
SDP
Supplier:  BEZWADA BIOMEDICAL LLC 71K004
Encompass_Preferred
Copolymer of PEG with absorbable polymers such as PLGA, PLA or PCL.The polymer is completely hydrolysable aand have controllable hydrolytic degradation profile. Polymers used in drug delivery and various biomedical applications
Catalog No. 50-190-4512