Learn More
Medchemexpress LLC PEG2000-DMPE | 99.7% | 2676.25 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11276850MG
Encompass_Preferred
PEG2000-DMPE can be used to synthesize a LNP. It enhances the entrapment efficiency depending on its increasing portion in the liposome. The optimal formulation for animal study is DMPC/PEG2000-DMPE/CH=50/5/45 at the weight ratio of drug/lipid=1/20.
  • Can be used to synthesize a LNP
  • Enhances entrapment efficiency
  • Optimal formulation for animal study is DMPC/PEG2000-DMPE/CH=50/5/45 at a weight ratio of drug/lipid=1/20
Catalog No. 50-004-37371