Learn More
Medchemexpress LLC PEG3-C4-OBn | 2381196-79-6 | 99.3% | 500 MG
SDP
Supplier:  Medchemexpress LLC HY130620500MG
Encompass_Preferred
PEG3-C4-OBn is a polyethylene glycol (PEG)-based PROTAC linker. It is utilized in the synthesis of PROTAC SGK3 degrader-1.
  • Polyethylene glycol (PEG)-based PROTAC linker
  • Utilized in the synthesis of PROTAC SGK3 degrader-1
  • Key component for protein degradation studies
  • Potential inhibitor for SGK3 related research
Catalog No. 50-003-40259