Learn More
Medchemexpress LLC PEG3-methylamine | 282551-10-4 | ≥98.0% | 163.21 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1401611G
Encompass_Preferred
PEG3-methylamine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
  • Liquid appearance
  • Yellow to brown color
  • Density: 1 g/cm3
Catalog No. 50-003-50520