Learn More
Medchemexpress LLC Peg4-aminooxy-mmaf | 1415246-35-3 | 99.8% | 923.19 g/mol | C47H82N6O12 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1289681MG
Encompass_Preferred
PEG4-aminooxy-MMAF is a drug-linker conjugate used in the preparation of antibody-drug conjugates (ADCs). It pairs the antitubulin payload monomethyl auristatin F (MMAF) with a noncleavable PEG4 aminooxy linker to allow stable conjugation and targeted delivery of the cytotoxic agent for research applications.
  • Stable noncleavable PEG4 aminooxy linker for antibody conjugation.
  • Contains the antitubulin payload monomethyl auristatin F (MMAF).
  • High purity as reported by supplier (99.83%).
  • Available in small milligram quantities for research use.
  • Intended for research use only; not validated for clinical use.
Catalog No. 50-002-03962