Learn More
Medchemexpress LLC Pegloxenatide | 2420483-82-3 | 90.00% | 500 UG
SDP
Supplier:  Medchemexpress LLC HYP3491500UG
Encompass_Preferred
Pegloxenatide is a long-acting glucagon-like peptide-1 receptor (GLP-1RA) agonist. It is used in the research of type 2 diabetes and its multiple complications, demonstrating effects such as lowering blood glucose and lipids, improving body weight, and providing anti-inflammatory, hepatoprotective, and cardioprotective activities.
  • Long-acting glucagon-like peptide-1 receptor (GLP-1RA) agonist
  • Lowers blood glucose and lipids
  • Improves body weight
  • Exhibits anti-inflammatory effects
  • Promotes wound healing
  • Protects the liver and heart
  • Activates PI3K/AKT pathway
  • Restores tube formation and migratory capacity
  • Reduces mitochondrial reactive oxygen species generation
Catalog No. 50-003-98737