Learn More
Hampton Research HR2-982-12/PEGRx 1 #12 (A12), 185 ml - 0.1 M BIS-TRIS propane pH 9.0, 20% v/v Polyethylene glycol monomethyl ether 550
SDP
Supplier:  Hampton Research HR298212
Encompass
HR2-982-12/PEGRx 1 #12 (A12), 185 ml - 0.1 M BIS-TRIS propane pH 9.0, 20% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.
Catalog No. 50-230-5