Learn More
Hampton Research HR2-982-14/PEGRx 1 #14 (B2), 185 ml - 0.1 M MES monohydrate pH 6.0, 20% v/v Jeffamine M-600 pH 7.0
SDP
Supplier:  Hampton Research HR298214
Encompass
HR2-982-14/PEGRx 1 #14 (B2), 185 ml - 0.1 M MES monohydrate pH 6.0, 20% v/v Jeffamine M-600 pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-230-7