Learn More
Hampton Research HR2-982-30/PEGRx 1 #30 (C6), 185 ml - 0.1 M HEPES pH 7.5, 12% w/v Polyethylene glycol 3,350
SDP
Supplier:  Hampton Research HR298230
Encompass
HR2-982-30/PEGRx 1 #30 (C6), 185 ml - 0.1 M HEPES pH 7.5, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-232-3