Learn More
Hampton Research HR2-982-40/PEGRx 1 #40 (D4), 185 ml - 0.1 M Citric acid pH 3.5, 28% w/v Polyethylene glycol 8,000
SDP
Supplier:  Hampton Research HR298240
Encompass
HR2-982-40/PEGRx 1 #40 (D4), 185 ml - 0.1 M Citric acid pH 3.5, 28% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-233-3