Learn More
Hampton Research HR2-982-46/PEGRx 1 #46 (D10), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 18% w/v Polyethylene glycol 20,000
SDP
Supplier:  Hampton Research HR298246
Encompass
HR2-982-46/PEGRx 1 #46 (D10), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 18% w/v Polyethylene glycol 20,000. Please note items from this supplier are non-returnable.
Catalog No. 50-233-9