Learn More
Hampton Research HR2-984-14/PEGRx 2 #14 (F02), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 20% w/v Polyethylene glycol 1,000
SDP
Supplier:  Hampton Research HR298414
Encompass
HR2-984-14/PEGRx 2 #14 (F02), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 20% w/v Polyethylene glycol 1,000. Please note items from this supplier are non-returnable.
Catalog No. 50-235-5