Learn More
Hampton Research HR2-984-15/PEGRx 2 #15 (F03), 185 ml - 0.1 M Sodium malonate pH 8.0, 0.1 M Tris pH 8.0, 30% w/v Polyethylene glycol 1,000
SDP
Supplier:  Hampton Research HR298415
Encompass
HR2-984-15/PEGRx 2 #15 (F03), 185 ml - 0.1 M Sodium malonate pH 8.0, 0.1 M Tris pH 8.0, 30% w/v Polyethylene glycol 1,000. Please note items from this supplier are non-returnable.
Catalog No. 50-235-6