Learn More
Hampton Research HR2-984-16/PEGRx 2 #16 (F04), 185 ml - 4% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.1 M Citric acid pH 3.5, 20% w/v Polyethylene glycol 1,500
SDP
Supplier:  Hampton Research HR298416
Encompass
HR2-984-16/PEGRx 2 #16 (F04), 185 ml - 4% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.1 M Citric acid pH 3.5, 20% w/v Polyethylene glycol 1,500. Please note items from this supplier are non-returnable.
Catalog No. 50-235-7