Learn More
Hampton Research HR2-984-22/PEGRx 2 #22 (F10), 185 ml - 0.2 M Ammonium citrate tribasic pH 7.0, 0.1 M Imidazole pH 7.0, 20% w/v Polyethylene glycol monomethyl ether 2,000
SDP
Supplier:  Hampton Research HR298422
Encompass
HR2-984-22/PEGRx 2 #22 (F10), 185 ml - 0.2 M Ammonium citrate tribasic pH 7.0, 0.1 M Imidazole pH 7.0, 20% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.
Catalog No. 50-236-3