Learn More
Hampton Research HR2-984-28/PEGRx 2 #28 (G04), 185 ml - 18% v/v 2-Propanol, 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 20% w/v Polyethylene glycol 4,000
SDP
Supplier:  Hampton Research HR298428
Encompass
HR2-984-28/PEGRx 2 #28 (G04), 185 ml - 18% v/v 2-Propanol, 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 20% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-236-9