Learn More
Enzo Life Sciences PEGylated protein ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI9002130001
Encompass_Preferred
Validated, Complete Assay for the Sensitive Detection of a Diverse Set of PEGs in Biological Samples. Alternative Name: Polyethylene glycol. Sensitivity: <1 ng/ml (range 1.75-225 ng/ml). Assay Time: 2 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: Microtiter Plate, PEG-BSA Assay Control, Biotinylated PEG, Assay Buffer 39, Conjugate, Wash Buffer 3, TMB Substrate, Stop Solution 2, Plate Sealer
Catalog No. NC0232106