Learn More
Creative Biomart Lipegfilgrastim, Pegylated GCSF
Supplier:  Creative Biomart THP0083
Encompass_Preferred
The product is a pegylated, recombinant granulocyte colony-stimulating factor (G-CSF) that was synthetized using a highly site-specific glycoPEGylation technology. The product is a covalent conjugate of Filgrastim with a single methoxy polyethylene glycol (PEG) molecule via a carbohydrate linker consisting of glycine, N-acetylneuraminic acid and N-acetylgalactosamine. The average molecular mass of the product comprises 18,798 Da for Filgrastim, 203 Da for GalNAc, 338 Da for glycylsialic acid and approximately 20,000 Da for PEG.
Catalog No. 50-208-4134