Learn More
MilliporeSigma Pellicon Mini Cassette Holder 1 ea
Supplier:  MilliporeSigma XX42PMINI
Encompass
Pellicon 2 Mini Holder,Qty/Pk: 1,Depth cm (in):14 (5.5),Width cm (in): 11.4 (4.5),Weight kg (lb): 5 (11), Maximum Inlet Pressure bar (psig): 7 (100)
Catalog No. NC9565846
Only null left
Add to Cart
Request a Quote