Learn More
Medchemexpress LLC Pelargonidin-3-O-glucoside chloride | 18466-51-8 | MFCD00017578 | 98.0% | 468.84 g/mol | C21H21ClO10 | 1 MG

Supplier: Medchemexpress LLC HYW0400451MG
Callistephin chloride (pelargonidin-3-O-glucoside chloride) is the chloride salt of the anthocyanin callistephin, supplied as a powdered analytical standard for biochemical and analytical work. It is used in antioxidant and reactive oxygen species (ROS) studies, and as a reference for chromatographic quantification.
- Analytical standard suitable for HPLC and quantitative assays.
- High purity (98.0%).
- Molecular weight 468.84 g/mol.
- Chemical formula C21H21ClO10.
- Available in small sizes appropriate for analytical workflows.
- Stable as a sealed powder when stored away from moisture and light at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.