Learn More
Ted Pella Inc PELCO Reverse, Fine Tweezer, 3X, 118mm, NM-SS, 0.10 x 0.15mm
SDP
Supplier:  Ted Pella Inc 5377NM
Encompass
High precision reverse action tweezers. Specially intended where a high precision self closing action is required, such as in microscopy and assembly applications. Extra fine tipped tweezers made of anti-acid / non-magnetic stainless steel.
Catalog No. NC0251179