Learn More
MSC Pelican Products Aircase with Divider, Layered Foam, HPX High Performance Resin, 14.5 in Wide
Supplier:  MSC 38471421
Encompass
This Aircase with Divider from Pelican Products offers robust protection for sensitive equipment. It features layered foam for customizable cushioning and is constructed from HPX High Performance Resin, ensuring durability and lightweight portability.
  • Business card holder
  • Automatic purge valve
  • Quiet rolling bearing wheels
  • Super-light proprietary design
  • Includes Trekpak insert
  • Padlockable
  • Meets IP67 standards for water and dust resistance
Catalog No. 50-005-28390