Learn More
Medchemexpress LLC Pelitinib | 257933-82-7 | 98.5% | 467.92 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY32718100MG
Encompass_Preferred
Pelitinib is an irreversible inhibitor of EGFR, also known to slightly inhibit Src, MEK/ERK, and ErbB2. It is intended for research use only.
  • Irreversible EGFR inhibitor
  • IC50 for EGFR is 38.5 nM
  • Slightly inhibits Src, MEK/ERK, and ErbB2
  • IC50s for Src, MEK/ERK, and ErbB2 are 282, 800, and 1255 nM, respectively
Catalog No. 50-004-03776