Learn More
Medchemexpress LLC Pellitorine | 18836-52-7 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN30971MG
Encompass_Preferred
Pellitorine is a bioactive natural amide compound that competitively antagonizes TRPV1 activation, thereby reducing pain signal transmission. It improves cognitive dysfunction by upregulating the BDNF-ERK1/2-CREB and Nrf2-HO-1 pathways, and exerts anti-inflammatory and anti-sepsis effects. Additionally, Pellitorine exhibits antithrombotic activity, inhibits lipid peroxidation, resists ferroptosis, and shows anti-cancer properties. It also acts as an insecticide against Aedes aegypti mosquito larvae.
  • Reduces pain by antagonizing TRPV1 activation.
  • Improves cognitive function via BDNF-ERK1/2-CREB and Nrf2-HO-1 pathways.
  • Exerts anti-inflammatory and anti-sepsis effects.
  • Provides antithrombotic effects.
  • Inhibits lipid peroxidation and resists ferroptosis.
  • Kills Aedes aegypti mosquito larvae.
  • Exhibits anti-cancer activity (leukemia and breast cancer).
  • Inhibits growth of certain bacteria.
Catalog No. 50-004-21307