Learn More
AdipoGen Pellitorine (1 mg)
SDP
Supplier:  AdipoGen AGCN20009M001
Encompass_Preferred
Chemical. CAS: 18836-52-7. Formula: C14H25NO. MW: 223.4. Synthetic. Originally isolated from roots of Anacyclus pryrethrum and fruits of Piper nigrum. Tingling-inducing agent. Excellent stable model compound for sensory studies. Exerts same profile as the unstable compound hydroxy-alpha-sanshool. Shows larvicidal, antimycobacterial and antituberculosis activity. alpha-Glucosidase inhibitor used in diabetes mellitus, cancer, infection and inflammatory research. ACAT (Acyl-CoA cholesteryl acyl transferase) inhibitor. Potential anti-cancer lead compound. Anti-thrombotic. Anti-septic. Antiprotozoal, antimalarial activity.
Catalog No. 50-205-3355