Learn More
Memmert Memmert Peltier Cooled Incubator IPP260Eco Plus. 260L - 115V, Stainless Steel Construction with SingleDISPLAY digital controller.
SDP
Supplier:  Memmert IPP260ECO PLUS 115V
Encompass_Preferred
Memmert Cooled-Incubators - this programmable model is the 115 volt, Peltier-technology seamlessly heats and cools, with TwinDISPLAY digital controller. Interior dimensions 640/800/468-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Atmocontrol Software, Manual, Quick Start Guide, Calibration Certificate and Power Cord - energy efficient, and precise! Working temperature range 0C (at least 20 below ambient) to +70C.
Catalog No. 50-225-4386
Only null left
Add to Cart
Request a Quote