Learn More
Memmert Memmert Peltier Cooled Incubator IPP30. 30L - 115V, Stainless Steel Construction with SingleDISPLAY digital controller.
SDP
Supplier:  Memmert IPP30 115V
Encompass_Preferred
Memmert Cooled-Incubators - this model is the 115 volt, Peltier-technology seamlessly heats and cools, with SingleDISPLAY digital controller. Interior dimensions 400/320/240-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Quick Start Guide, Calibration Certificate and Power Cord - energy efficient, and precise! Working temperature range 0C (at least 20 below ambient) to +70C.
Catalog No. 50-225-4379