Learn More
Medchemexpress LLC HY-P9902 5mg , Pembrolizumab CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99025MG
Encompass_Preferred
HY-P9902 5mg Pembrolizumab CAS: Pembrolizumab (MK-3475) is a humanized IgG4 antibody inhibiting the programmed cell death 1 (PD-1) receptor, used in cancer immunotherapy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8983