Learn More
Genscript Corporation Antibody Kit Other Products Immunodetection 1/EA
Supplier:  Genscript Corporation L00971
Encompass
GenScript Pembrolizumab Immunogenicity Kit (Bridging ELISA) is used for detection of anti-Pembrolizumab antibody in serum and plasma samples. It is a validated bridging immunoassay based on the FDA EMA and NMPA Immunogenicity Guidelines. The kit has been proved to be highly sensitive specific and free from matrix effect in the validation study.
Catalog No. 50-252-9974