Learn More
Apexbio Technology LLC Pemetrexed disodium hemipenta hydrate 357166-30-4 200mg
SDP
Supplier:  Apexbio Technology LLC A3707200
Encompass_Preferred
Pemetrexed disodium hemipenta hydrate (CAS 357166-30-4) is a multitargeted antifolate that inhibits key enzymes in folate metabolism including thymidylate synthase (TS) dihydrofolate reductase (DHFR) and glycinamide ribonucleotide formyltransferase (GARFT) with Ki values for its pentaglutamate form of 1 3 nM 7 2 nM and 65 nM respectively Compared to the monoglutamate form the pentaglutamate exhibits enhanced inhibitory potency particularly against TS and GARFT Pemetrexed is approved for the treatment of advanced non-squamous non-small cell lung cancer In preclinical models it additionally modulates renal uptake of radiolabeled folates These features support its utilization in studies of folate pathway inhibition and cancer therapy
Catalog No. 50-000-95476