Learn More
Selleck Chemical LLC Pemigatinib S0088-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S008810MM/1ML
Encompass_Preferred
Pemigatinib is an orally active and selective inhibitor of FGFR with IC50 of 0 4 nM 0 5 nM 1 2 nM and 30 nM for FGFR1 FGFR2 FGFR3 and FGFR4 respectively Pemigatinib has the potential for cholangiocarcinoma
Catalog No. 50-000-39390