Learn More
Medchemexpress LLC Pen-N3 (PDZ peptide) | 98.8% | 1071.22 | 5 MG

Supplier: Medchemexpress LLC HYP101645MG
Pen-N3 is a PDZ peptide that inhibits Wnt/β-catenin signaling by recognizing dishevelled protein.
- Molecular weight: 1071.22
- Formula: C51H78N10O15
- Appearance: Solid
- Color: White to off-white
- Sequence: Glu-Ile-Val-Leu-Trp-Ser-Asp-Ile-Pro
- Sequence shortening: EIVLWSDIP
- Solubility in DMSO: 100 mg/mL (93.35 mM; needs ultrasonic; hygroscopic DMSO has a significant impact on product solubility, please use newly opened DMSO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.