Learn More
Quality Biological Inc Penicillin/Streptomycin Mixture 10mL 10/PK
SDP
Supplier:  Quality Biological Inc 120095671
Encompass_Preferred
Penicillin-Streptomycin acts in two ways. Penicillin inhibits bacterial cell wall synthesis. While streptomycin inhibits prokaryote protein synthesis by preventing the transition from initiation complex to chain-elongating ribosome causing miscoding.100X concentration of mixture of penicillin and streptomycin Manufactured in an ISO 90012015 certified facility.
Catalog No. 50-220-9762