Learn More
Medchemexpress LLC Penao | 1192411-43-0 | MFCD25976838 | 96.7% | 390.29 | C13H19AsN2O5S | 10 MG

Supplier: Medchemexpress LLC HY1638610MG
PENAO is a small-molecule arsenical mitochondrial toxin that inactivates tumor cell mitochondria by targeting the inner-membrane adenine nucleotide transferase (ANT). It is supplied as a research reagent in milligram quantities for in vitro studies; the compound contains arsenic and must be handled per the safety data sheet (SDS).
- Potent tumor cell mitochondrial toxin that disrupts mitochondrial function.
- Mechanism: targets inner-membrane adenine nucleotide transferase (ANT).
- Available in small-milligram quantities suitable for laboratory research.
- Purity approximately 96.7% as provided by the manufacturer.
- Contains arsenic; follow the safety data sheet (SDS) for handling and disposal.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.