Learn More
Cayman Chemical PendImethalIn 100mg
SDP
Supplier:  Cayman Chemical 34557100 MG
Encompass_Preferred
A dinitroaniline herbicide; controls johnsongrass and increases seed cotton yield from 0.56 to 1.12 kg/ha; induces the production of ROS and genotoxicity in isolated human peripheral lymphocytes from 25-500 µM; increases renal and hepatic protein carbonylation, lipid peroxidation, necrosis, leukocyte infiltration, and formation of pyknotic nuclei in rats at 62.5, 125, and 250 mg/kg
Catalog No. 50-294-7191