Learn More
Medchemexpress LLC Penicillic acid | 90-65-3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN677710MM/1ML
Encompass_Preferred
Penicillic acid is a polyketide mycotoxin produced by Aspergillus and Penicillium species. It exhibits cytotoxicity in rat alveolar macrophages in vitro and inhibits Fas ligand-induced apoptosis by blocking self-processing of caspase-8. This compound is provided as a white to off-white solid with a molecular formula of C8H10O4 and a molecular weight of 170.16.
  • Causes significant chromium release in alveolar macrophages at 0.1 mM.
  • Decreases ATP levels in alveolar macrophages at 1 mM.
  • Inhibits protein synthesis in alveolar macrophages at 0.3 mM.
  • Dose-dependently inhibits DNA fragmentation induced by Jo2 in A2o cells.
  • Reverses the reduction in cell viability of A2o cells exposed to Jo2.
  • Prevents caspase-8 processing in Raji cells treated with higher concentrations of FasL.
Catalog No. 50-004-18272