Learn More
Medchemexpress LLC Penicillin G-d7 (potassium) | 352323-25-2 | 2.5 MG
SDP
Supplier:  Medchemexpress LLC HYN7139S12.5MG
Encompass_Preferred
Penicillin G-d7 (potassium), also known as Benzylpenicillin-d7 potassium salt, is a deuterium-labeled variant of Penicillin G potassium salt. This compound is intended for research use only and is not sold to patients. It can be utilized as a tracer and as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-17815