Learn More
Alkali Scientific Penicillin G Potassium Salt, 25 G
Supplier: Alkali Scientific ENI002CP
Penicillin G Potassium Salt, 25 g, is an antibiotic powder used to treat infections caused by gram-positive bacteria. The potassium form of Penicillin G is commonly employed in laboratory settings for testing antibacterial properties and researching microbial resistance. Penicillin G works by targeting bacterial cell wall synthesis, making it effective in disrupting bacterial growth and reproduction. This formulation is widely used in microbiological studies to investigate the effects of Penicillin on bacterial cultures, evaluate susceptibility, and assess resistance mechanisms. The 25 g package is ideal for preparing antimicrobial testing solutions or for larger-scale studies in pharmaceutical research and clinical applications.