Learn More
Sigma Aldrich Fine Chemicals Biosciences Penicillin-Streptomycin, 20 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P433320ML
Encompass
Penicillin-Streptomycin is used to supplement cell culture media to control bacterial contamination. It is recommended for use in cell culture applications at 10 mL/L.
Catalog No. NC0855654